2-Bromo-1,1-difluoroethylene 98%
Supplier: Apollo Scientific
Danger
PC1390-25G PC1390-100G PC1390-5G
APOSPC1390-25GEA 368 EUR
APOSPC1390-25G APOSPC1390-100G APOSPC1390-5G
2-Bromo-1,1-difluoroethylene 98%
2-Bromo-1,1-difluoroethylene
Formula: C₂HBrF₂
Boiling Pt: 6.1 °C
Melting Pt: –115 °C
Density: 1.817 g/cm³ (25 °C)
Flash Pt: <23 °C
Storage Temperature: Ambient
CAS Number: 359-08-0
UN: 3161
ADR: 2.1,