3-Fluoro-2-nitrobenzoic acid 98+%
Supplier: Apollo Scientific
Warning
PC1396-10G PC1396-1G PC1396-5G
APOSPC1396-10GEA 195 EUR
APOSPC1396-10G APOSPC1396-1G APOSPC1396-5G
3-Fluoro-2-nitrobenzoic acid 98+%
3-Fluoro-2-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 192…195 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07368342
CAS Number: 1000339-51-4