3'-Bromo-4'-fluoroacetophenone 97%
Supplier: Apollo Scientific
Warning
PC1396C-10G PC1396C-25G
APOSPC1396C-10GEA 24.8 EUR
APOSPC1396C-10G APOSPC1396C-25G
3'-Bromo-4'-fluoroacetophenone 97%
3'-Bromo-4'-fluoroacetophenone
Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 150 °C/12mmHg
Melting Pt: 51…57 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042466
CAS Number: 1007-15-4
EINECS: 213-750-0