5-Bromo-2-fluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC1399G-100G PC1399G-25G PC1399G-500G
APOSPC1399G-100GEA 30.6 EUR
APOSPC1399G-100G APOSPC1399G-25G APOSPC1399G-500G
5-Bromo-2-fluorobenzaldehyde 98%
5-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
Boiling Pt: 230 °C
Melting Pt: 23 °C
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00070755
CAS Number: 93777-26-5
EINECS: 298-056-6