4-Fluoropyridine hydrochloride 95%
Supplier: Apollo Scientific
Warning
PC1406-1G PC1406-5G PC1406-250MG
APOSPC1406-1GEA 168 EUR
APOSPC1406-1G APOSPC1406-5G APOSPC1406-250MG
4-Fluoropyridine hydrochloride 95%
4-Fluoropyridine hydrochloride
Formula: C₅H₅ClFN
Melting Pt: 110 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00234055
CAS Number: 39160-31-1