1-Bromo-3-fluorobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromofluorobenzene
PC1410-500G PC1410-100G
APOSPC1410-500GEA 87 EUR
APOSPC1410-500G APOSPC1410-100G
1-Bromo-3-fluorobenzene 99%
1-Bromo-3-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 148…150 °C
Melting Pt: -8
Density: 1.567 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000326
CAS Number: 1073-06-9
EINECS: 214-023-0
UN: 1993
ADR: 3 III