1-Bromo-4-fluorobenzene 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromofluorobenzene
PC1420-100G
PC1420-1KG
PC1420-500G
APOSPC1420-100GEA
26.1
EUR
APOSPC1420-100G
APOSPC1420-1KG
APOSPC1420-500G
1-Bromo-4-fluorobenzene 99%
1-Bromo-4-fluorobenzene
Formula:
C₆H₄BrF MW: 175 g/mol Boiling Pt: 152…155 °C Melting Pt: –17 °C Density: 1.593 g/cm³ (25 °C) Flash Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000342 CAS Number: 460-00-4 EINECS: 207-300-2 UN: 1993 ADR: 3 III |