3-Bromo-4-fluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC1420H-100G PC1420H-25G PC1420H-500G
APOSPC1420H-100GEA 92.8 EUR
APOSPC1420H-100G APOSPC1420H-25G APOSPC1420H-500G
3-Bromo-4-fluorobenzonitrile 98%
3-Bromo-4-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 134…136 °C/33mmHg
Melting Pt: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00055432
CAS Number: 79630-23-2
EINECS: 279-200-7
UN: 3439
ADR: 6.1 III