2-Bromo-α,α,α,5-tetrafluorotoluene 97%
Supplier: Apollo Scientific
Warning
PC1421-100G PC1421-500G PC1421-1KG
APOSPC1421-100GEA 42.2 EUR
APOSPC1421-100G APOSPC1421-500G APOSPC1421-1KG
2-Bromo-α,α,α,5-tetrafluorotoluene 97%
2-Bromo-α,α,α,5-tetrafluorotoluene
Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 136…143 °C
Density: 1.695 g/cm³ (25 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00040937
CAS Number: 40161-55-5