3-Bromo-α,α,α,4-tetrafluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-1-fluoro-4-(trifluoromethyl)benzene, 3-Bromo-4-fluorobenzotrifluoride, 1-Bromo-2-fluoro-5-(trifluoromethyl)benzene
PC1422-500G PC1422-100G PC1422-25G
APOSPC1422-500GEA 142 EUR
APOSPC1422-500G APOSPC1422-100G APOSPC1422-25G
3-Bromo-α,α,α,4-tetrafluorotoluene 98%
3-Bromo-α,α,α,4-tetrafluorotoluene
Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 148…149 °C
Density: 1.706 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00040938
CAS Number: 68322-84-9