5-Bromo-α,α,α,2-tetrafluorotoluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-fluorobenzotrifluoride, 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene
PC1423D-25G PC1423D-100G PC1423D-500G
APOSPC1423D-25GEA 24.8 EUR
APOSPC1423D-25G APOSPC1423D-100G APOSPC1423D-500G
5-Bromo-α,α,α,2-tetrafluorotoluene 97%
5-Bromo-α,α,α,2-tetrafluorotoluene
Formula: C₇H₃BrF₄
Boiling Pt: 79…80 °C/50mmHg
Density: 1.72 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042493
CAS Number: 393-37-3