1-Bromo-3-fluoro-4-iodobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Fluoro-4-iodobromobenzene, 4-Bromo-2-fluoro-1-iodobenzene
PC1426F-5G PC1426F-25G PC1426F-100G PC1426F-500G
APOSPC1426F-5GEA 0 EUR
APOSPC1426F-5G APOSPC1426F-25G APOSPC1426F-100G APOSPC1426F-500G
1-Bromo-3-fluoro-4-iodobenzene 97%
1-Bromo-3-fluoro-4-iodobenzene
Formula: C₆H₃BrFI
Boiling Pt: 48…51 °C
Melting Pt: 48…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00010608
CAS Number: 105931-73-5