1-Bromo-4-fluoronaphthalene
Supplier: Apollo Scientific
Warning
PC1426R-100G PC1426R-25G
APOSPC1426R-100GEA 114 EUR
APOSPC1426R-100G APOSPC1426R-25G
1-Bromo-4-fluoronaphthalene
1-Bromo-4-fluoronaphthalene
Formula: C₁₀H₆BrF
MW: 225.06 g/mol
Melting Pt: 33…37 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051473
CAS Number: 341-41-3
EINECS: 206-434-9