1-Bromo-4-fluoro-3-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1-fluoro-2-nitrobenzene, 5-Bromo-2-fluoronitrobenzene
PC1428D-100G PC1428D-25G PC1428D-500G
APOSPC1428D-100GEA 26.1 EUR
APOSPC1428D-100G APOSPC1428D-25G APOSPC1428D-500G
1-Bromo-4-fluoro-3-nitrobenzene 98%
1-Bromo-4-fluoro-3-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 240…241 °C
Melting Pt: 18…19 °C
Density: 1.786 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00129165
CAS Number: 364-73-8