4-Bromo-2-fluoro-6-nitrophenol 97%
Supplier: Apollo Scientific
Warning
PC1428G-25G PC1428G-5G
APOSPC1428G-25GEA 79.9 EUR
APOSPC1428G-25G APOSPC1428G-5G
4-Bromo-2-fluoro-6-nitrophenol 97%
4-Bromo-2-fluoro-6-nitrophenol
Formula: C₆H₃BrFNO₃
MW: 236.00 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00013372
CAS Number: 320-76-3