2-Bromo-5-fluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-fluoro-2-methylbenzene
PC1430A-100G PC1430A-25G
APOSPC1430A-100GEA 37.7 EUR
APOSPC1430A-100G APOSPC1430A-25G
2-Bromo-5-fluorotoluene 98%
2-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 176…177 °C
Density: 1.495 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3 III