1,3-Dichloro-2-fluoro-4-nitrobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dichloro-3-fluoronitrobenzene
PC1434-1G PC1434-5G PC1434-25G
APOSPC1434-1GEA 21.9 EUR
APOSPC1434-1G APOSPC1434-5G APOSPC1434-25G
1,3-Dichloro-2-fluoro-4-nitrobenzene
1,3-Dichloro-2-fluoro-4-nitrobenzene
Formula: C₆H₂Cl₂FNO₂
MW: 209.9900 g/mol
Boiling Pt: 88 °C/1mmHg
Density: 1.6 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042240
CAS Number: 393-79-3