3-Bromo-4-fluorotoluene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2-fluoro-5-methylbenzene
PC1440-100G PC1440-250G PC1440-25G
APOSPC1440-100GEA 58 EUR
APOSPC1440-100G APOSPC1440-250G APOSPC1440-25G
3-Bromo-4-fluorotoluene 99%
3-Bromo-4-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 168…170 °C
Density: 1.507 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00040827
CAS Number: 452-62-0