4-Bromo-3-fluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2-fluoro-4-methylbenzene
PC1451-100G PC1451-25G
APOSPC1451-100GEA 72.5 EUR
APOSPC1451-100G APOSPC1451-25G
4-Bromo-3-fluorotoluene 98%
4-Bromo-3-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 95 °C/50mmHg
Density: 1.494 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00040940
CAS Number: 452-74-4
UN: 1993
ADR: 3,III