5-Bromo-2,3-difluorophenol 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3,4-difluoro-5-hydroxybenzene
PC1452-100G PC1452-25G PC1452-500G
APOSPC1452-100GEA 104 EUR
APOSPC1452-100G APOSPC1452-25G APOSPC1452-500G
5-Bromo-2,3-difluorophenol 99%
5-Bromo-2,3-difluorophenol
Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Density: 1.8 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631350
CAS Number: 186590-26-1