3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-carbonitrile 95%
Supplier: Apollo Scientific
PC1455-1G PC1455-500MG
APOSPC1455-1GEA 328 EUR
APOSPC1455-1G APOSPC1455-500MG
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-carbonitrile 95%
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-carbonitrile
Formula: C₁₄H₁₀FNO
MW: 227.24 g/mol
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD06797371
CAS Number: 123864-93-7