5-Fluoro-2-methoxybenzonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-Cyano-4-fluoroanisole
PC14564-1G PC14564-25G PC14564-5G
APOSPC14564-1GEA 21.9 EUR
APOSPC14564-1G APOSPC14564-25G APOSPC14564-5G
5-Fluoro-2-methoxybenzonitrile
5-Fluoro-2-methoxybenzonitrile
Formula: C₈H₆FNO
MW: 151.14 g/mol
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD02683503
CAS Number: 189628-38-4