4'-Fluoro-2-hydroxyacetophenone 95%
Supplier: Apollo Scientific
Warning
PC1458-100G PC1458-25G PC1458-5G
APOSPC1458-100GEA 169 EUR
APOSPC1458-100G APOSPC1458-25G APOSPC1458-5G
4'-Fluoro-2-hydroxyacetophenone 95%
4'-Fluoro-2-hydroxyacetophenone
Formula: C₈H₇FO₂
Melting Pt: 113…117 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02679035
CAS Number: 403-31-6