(1-Bromo)perfluoro-2,5,8-trioxadodecane
Supplier: Apollo Scientific
Warning
Synonyms: Perfluoro-2,5-8-trioxadodecyl bromide
PC1473-25G PC1473-5G
APOSPC1473-25GEA 301 EUR
APOSPC1473-25G APOSPC1473-5G
(1-Bromo)perfluoro-2,5,8-trioxadodecane
(1-Bromo)perfluoro-2,5,8-trioxadodecane
Formula: C₉BrF₁₉O₃
MW: 596.98 g/mol
Boiling Pt: 60 °C/25mmHg
Storage Temperature: Ambient
MDL Number: MFCD03094130
CAS Number: 330562-47-5