6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 95+%
Supplier: Apollo Scientific
Danger
PC1476-1G PC1476-5G
APOSPC1476-1GEA 77 EUR
APOSPC1476-1G APOSPC1476-5G
6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 95+%
6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅BrF₃NO
MW: 292.06 g/mol
Melting Pt: >290 °C
Storage Temperature: Ambient
MDL Number: MFCD00153078
CAS Number: 1701-22-0
UN: 2811
ADR: 6.1 III