1-Bromotetrafluoro-2-iodoethane 97%
Supplier: Apollo Scientific
Warning
PC1480-25G PC1480-2G PC1480-5G
APOSPC1480-25GEA 271 EUR
APOSPC1480-25G APOSPC1480-2G APOSPC1480-5G
1-Bromotetrafluoro-2-iodoethane 97%
1-Bromotetrafluoro-2-iodoethane
Formula: BrF₂CCF₂I
MW: 306.82 g/mol
Boiling Pt: 78 °C
Density: 1.648 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042136
CAS Number: 421-70-5
EINECS: 207-008-5