Ethyl-4-fluoro-3-nitrobenzoate
Supplier: Apollo Scientific
Warning
PC1485-100G PC1485-25G PC1485-5G
APOSPC1485-100GEA 139 EUR
APOSPC1485-100G APOSPC1485-25G APOSPC1485-5G
Ethyl-4-fluoro-3-nitrobenzoate
Ethyl-4-fluoro-3-nitrobenzoate
Formula: C₉H₈FNO₄
Boiling Pt: 128…130 °C/0.5mmHg
Melting Pt: 46…47 °C
Storage Temperature: Ambient
MDL Number: MFCD03428515
CAS Number: 367-80-6