1,3,5-Trifluoro-2,4,6-triiodobenzene
Supplier: Apollo Scientific
Warning
PC1493-25G
APOSPC1493-25GEA 121 EUR
APOSPC1493-25G
1,3,5-Trifluoro-2,4,6-triiodobenzene
1,3,5-Trifluoro-2,4,6-triiodobenzene
Formula: C₆F₃I₃
MW: 509.77 g/mol
Melting Pt: 151…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06248899
CAS Number: 84322-56-5