3-Fluoro-2-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Amino-6-fluorobenzotrifluoride
PC1495-1G PC1495-5G PC1495-25G
APOSPC1495-1GEA 42.2 EUR
APOSPC1495-1G APOSPC1495-5G APOSPC1495-25G
3-Fluoro-2-(trifluoromethyl)aniline 98%
3-Fluoro-2-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
MW: 179.12 g/mol
Flash Pt: <90 °C
Storage Temperature: Ambient
MDL Number: MFCD06658260
CAS Number: 123973-22-8
UN: 2810
ADR: 6.1,III