4-Bromo-3-nitrobenzotrifluoride 99%
Supplier: Apollo Scientific
Warning
PC1500-100G PC1500-25G
APOSPC1500-100GEA 58 EUR
APOSPC1500-100G APOSPC1500-25G
4-Bromo-3-nitrobenzotrifluoride 99%
4-Bromo-3-nitrobenzotrifluoride
Formula: C₇H₃BrF₃NO₂
MW: 270.01 g/mol
Boiling Pt: 90…92 °C/5mmHg
Density: 1.763 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014685
CAS Number: 349-03-1