5-Fluoro-2-methylbenzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC1523-25G PC1523-5G PC1523-100G
APOSPC1523-25GEA 55.1 EUR
APOSPC1523-25G APOSPC1523-5G APOSPC1523-100G
5-Fluoro-2-methylbenzaldehyde 95%
5-Fluoro-2-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 99 °C/30mmHg
Density: 1.145 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD02261763
CAS Number: 22062-53-9