1-Bromo-4-(trifluoromethylthio)benzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Trifluoromethylthio)bromobenzene, 4-Bromophenyl trifluoromethyl sulphide
PC1536-10G PC1536-50G PC1536-1G
APOSPC1536-10GEA 87 EUR
APOSPC1536-10G APOSPC1536-50G APOSPC1536-1G
1-Bromo-4-(trifluoromethylthio)benzene 97%
1-Bromo-4-(trifluoromethylthio)benzene
Formula: C₇H₄BrF₃S
MW: 257.07 g/mol
Boiling Pt: 81 °C/15mmHg
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00040840
CAS Number: 333-47-1