1-Bromo-2,3,4,5-tetrafluorobenzene 98%
Supplier: Apollo Scientific
Warning
PC1540-1G PC1540-5G
APOSPC1540-1GEA 98.9 EUR
APOSPC1540-1G APOSPC1540-5G
1-Bromo-2,3,4,5-tetrafluorobenzene 98%
1-Bromo-2,3,4,5-tetrafluorobenzene
Formula: C₆HBrF₄
MW: 228.97 g/mol
Boiling Pt: 48…55 °C/25mmHg
Density: 1.846 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00017900
CAS Number: 1074-91-5
EINECS: 214-048-7
UN: 1993
ADR: 3,III