Methyl-4-(trifluoromethylthio)benzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-[(trifluoromethyl)sulphanyl]benzoate
PC1546-1G PC1546-25G PC1546-5G
APOSPC1546-1GEA 58 EUR
APOSPC1546-1G APOSPC1546-25G APOSPC1546-5G
Methyl-4-(trifluoromethylthio)benzoate
Methyl 4-(trifluoromethylthio)benzoate
Formula: C₉H₇F₃O₂S
Boiling Pt: 117 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD06203725
CAS Number: 88489-60-5