4-Bromo-3,3,4,4-tetrafluoro-1-butanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-3,3,4,4-tetrafluorobutan-1-ol
PC1561F-25G PC1561F-5G PC1561F-100G
APOSPC1561F-25GEA 82.8 EUR
APOSPC1561F-25G APOSPC1561F-5G APOSPC1561F-100G
4-Bromo-3,3,4,4-tetrafluoro-1-butanol 97%
4-Bromo-3,3,4,4-tetrafluoro-1-butanol
Formula: C₄H₅BrF₄O
MW: 224.99 g/mol
Boiling Pt: 148 °C/30mmHg
Density: 1.66 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077467
CAS Number: 234443-21-1