2-Fluoro-3-iodopyridine
Supplier: Apollo Scientific
Danger
PC1564-5G
APOSPC1564-5GEA 29 EUR
APOSPC1564-5G
2-Fluoro-3-iodopyridine
2-Fluoro-3-iodopyridine
Formula: C₅H₃FIN
MW: 222.99 g/mol
Boiling Pt: 97…99 °C/17mmHg
Melting Pt: 42…48 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD03095287
CAS Number: 113975-22-7