1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)benzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-alpha,alpha,beta,beta-tetrafluorophenetole
PC1567-25G PC1567-5G PC1567-100G
APOSPC1567-25GEA 194 EUR
APOSPC1567-25G APOSPC1567-5G APOSPC1567-100G
1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)benzene
1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)benzene
Formula: C₈H₅BrF₄O
MW: 273.02 g/mol
Boiling Pt: 195…196 °C
Density: 1.628 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00042190
CAS Number: 68834-05-9
EINECS: 272-431-4