2-Bromo-2,3,3,3-tetrafluoropropionyl chloride 96%
Supplier: Apollo Scientific
Danger
PC1567P-250MG
APOSPC1567P-250MGEA 58 EUR
APOSPC1567P-250MG
2-Bromo-2,3,3,3-tetrafluoropropionyl chloride 96%
2-Bromo-2,3,3,3-tetrafluoropropionyl chloride
Formula: C₃BrClF₄O
MW: 243.39 g/mol
Boiling Pt: 67…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153723
CAS Number: 6066-45-1
UN: 3265
ADR: 8,II