1H,1H-Perfluoro-3,6,9-trioxadecan-1-ol 98%
Supplier: Apollo Scientific
Warning
PC1578-25G PC1578-5G
APOSPC1578-25GEA 102 EUR
APOSPC1578-25G APOSPC1578-5G
1H,1H-Perfluoro-3,6,9-trioxadecan-1-ol 98%
1H,1H-Perfluoro-3,6,9-trioxadecan-1-ol
Formula: C₇H₃F₁₃O₄
MW: 398.08 g/mol
Boiling Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00156026
CAS Number: 147492-57-7