Bromotrifluoroethylene 98%

Supplier: Apollo Scientific
Danger

PC1590-25G
APOSPC1590-25GEA 607 EUR
APOSPC1590-25G
Bromotrifluoroethylene 98%
Bromotrifluoroethylene

  • Pyrophoric

Formula: C₂BrF₃
Boiling Pt: –2 to 0 °C
Density: 1.86 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 598-73-2
UN: 2419
ADR: 2.1,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR