4-Bromo-1-iodo-2-(trifluoromethoxy)benzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-(trifluoromethoxy)iodobenzene
PC1593T-25G PC1593T-5G PC1593T-100G
APOSPC1593T-25GEA 36.4 EUR
APOSPC1593T-25G APOSPC1593T-5G APOSPC1593T-100G
4-Bromo-1-iodo-2-(trifluoromethoxy)benzene 97%
4-Bromo-1-iodo-2-(trifluoromethoxy)benzene
Formula: C₇H₃BrF₃IO
Boiling Pt: 112…114 °C/15mmHg
Melting Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00179334
CAS Number: 175278-12-3