2-Bromo-5-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Danger
PC1598G-5G PC1598G-25G PC1598G-1G
APOSPC1598G-5GEA 24.8 EUR
APOSPC1598G-5G APOSPC1598G-25G APOSPC1598G-1G
2-Bromo-5-(trifluoromethyl)pyridine 97%
2-Bromo-5-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 78…81 °C/30mmHg
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00153086
CAS Number: 50488-42-1
UN: 2811
ADR: 6.1,III