1-Bromo-3,3,3-trifluoro-2-propanol 96%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-1,1,1-trifluoropropan-2-ol, 3-Bromo-1,1,1-trifluoro-2-propanol
PC1599-1G PC1599-5G
APOSPC1599-1GEA 43.5 EUR
APOSPC1599-1G APOSPC1599-5G
1-Bromo-3,3,3-trifluoro-2-propanol 96%
1-Bromo-3,3,3-trifluoro-2-propanol
Precursor for 2-(Trifluoromethyl)oxirane
Formula: BrH₂CCH(OH)CF₃
Boiling Pt: 124…126 °C
Density: 1.861 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00040945
CAS Number: 431-34-5
UN: 1987
ADR: 3,III