3-Bromo-1,1,1-trifluoroacetone 97%
Supplier: Apollo Scientific
Danger
PC1600-25G PC1600-100G
APOSPC1600-25GEA 46.4 EUR
APOSPC1600-25G APOSPC1600-100G
3-Bromo-1,1,1-trifluoroacetone 97%
3-Bromo-1,1,1-trifluoroacetone
Formula: BrH₂CC(O)CF₃
Boiling Pt: 85…87 °C
Melting Pt: –80 °C
Density: 1.839 g/cm³ (25 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00039237
CAS Number: 431-35-6
EINECS: 207-071-9
UN: 2924
ADR: 3+8,II