3-(4-Fluorophenylcarbamoyl)phenylboronic acid pinacol ester 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-[(4-Fluorophenyl)aminocarbonyl]benzeneboronic acid, pinacol ester
PC1606-1G PC1606-5G
APOSPC1606-1GEA 215 EUR
APOSPC1606-1G APOSPC1606-5G
3-(4-Fluorophenylcarbamoyl)phenylboronic acid pinacol ester 95%
3-(4-Fluorophenylcarbamoyl)phenylboronic acid pinacol ester
Formula: C₁₉H₂₁BFNO₃
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD06659930
CAS Number: 850567-58-7