n-Butyl trifluoroacetate 97%
Supplier: Apollo Scientific
Danger
PC1614-100G PC1614-25G
APOSPC1614-100GEA 158 EUR
APOSPC1614-100G APOSPC1614-25G
n-Butyl trifluoroacetate 97%
n-Butyl trifluoroacetate
Formula: C₆H₉F₃O₂
MW: 170.13 g/mol
Boiling Pt: 72 °C
Density: 1.098 g/cm³ (25 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00039234
CAS Number: 367-64-6
EINECS: 206-699-0
UN: 2924
ADR: 3+8 II