2-Chloro-4-fluoroanisole 98%
Supplier: Apollo Scientific
Warning
PC1623-100G PC1623-25G PC1623-5G
APOSPC1623-100GEA 295 EUR
APOSPC1623-100G APOSPC1623-25G APOSPC1623-5G
2-Chloro-4-fluoroanisole 98%
2-Chloro-4-fluoroanisole
Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 197…200 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00672965
CAS Number: 2267-25-6