Cesium fluoride, anhydrous 99,5%
Supplier: Apollo Scientific
Danger
PC1630-100G PC1630-25G PC1630-500G
APOSPC1630-100GEA 68.3 EUR
APOSPC1630-100G APOSPC1630-25G APOSPC1630-500G
Cesium fluoride, anhydrous 99,5%
Cesium fluoride
Formula: CsF
Boiling Pt: 1251 °C
Melting Pt: 682 °C
Density: 4.115 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 3288
ADR: 6.1,III