N-(2,4-Difluorophenyl)-2-chloroacetamide 97%
Supplier: Apollo Scientific
Warning
PC1686-5G PC1686-1G PC1686-10G
APOSPC1686-5GEA 62.5 EUR
APOSPC1686-5G APOSPC1686-1G APOSPC1686-10G
N-(2,4-Difluorophenyl)-2-chloroacetamide 97%
N-(2,4-Difluorophenyl)-2-chloroacetamide
Formula: C₈H₆ClF₂NO
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00077473
CAS Number: 96980-65-3