2-Chloro-4'-fluoroacetanilide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-N-(4-fluorophenyl)acetamide, N-(Chloroacetyl)-4-fluoroaniline
PC1688F-100G PC1688F-25G PC1688F-5G
APOSPC1688F-100GEA 192 EUR
APOSPC1688F-100G APOSPC1688F-25G APOSPC1688F-5G
2-Chloro-4'-fluoroacetanilide 97%
2-Chloro-4'-fluoroacetanilide
Formula: C₈H₇ClFNO
MW: 187.6 g/mol
Melting Pt: 131…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00015730
CAS Number: 351-04-2